siRNA Details
| Gene Symbol: Hps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176785.3 | NP_789742.2 | 1 | 2254 | ||
| GenBank mRNA | AF536239.1 | - | 1 | 2199 | ||
| AK030582.1 | - | 1 | 2225 | |||
| AK040849.1 | - | 1 | 2201 | |||
| AK159588.1 | - | 1 | 2220 | |||
| AK172058.1 | - | 2 | 2005 | |||
| AK216285.1 | - | 1 | 17 | |||
| BC034055.1 | - | 1 | 2200 | 3673655 | ||

Hps6
Genomic Map