siRNA Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293660.1 | NP_001280589.1 | 8 | 865 | ||
| NM_011307.2 | NP_035437.1 | 10 | 1703 | |||
| GenBank mRNA | AK052704.1 | - | 10 | 1704 | ||
| AK083342.1 | - | 10 | 1644 | |||
| AK160926.1 | - | 8 | 866 | |||
| BC037092.1 | - | 9 | 1578 | |||
| BC040808.1 | - | 10 | 1659 | 4038778 | ||
| BC051659.1 | - | 10 | 1626 | |||
| BC066220.1 | - | 10 | 1626 | |||
| U22015.1 | - | 6 | 1155 | |||

Uimc1
Genomic Map