siRNA Details
| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009108.2 | NP_033134.2 | 3, 4 | 656 | ||
| GenBank mRNA | AK132255.1 | - | 3, 4 | 648 | ||
| BC015261.1 | - | 3, 4 | 627 | 4241705 | ||
| U09416.1 | - | 3, 4 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr1h4| Gene Symbol: Nr1h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009108.2 | NP_033134.2 | 3, 4 | 656 | ||
| GenBank mRNA | AK132255.1 | - | 3, 4 | 648 | ||
| BC015261.1 | - | 3, 4 | 627 | 4241705 | ||
| U09416.1 | - | 3, 4 | 625 | |||