siRNA Details
| Gene Symbol: Ryk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042607.1 | NP_001036072.1 | 4 | 589 | ||
| NM_001284258.1 | NP_001271187.1 | 3 | 467 | |||
| NM_013649.3 | NP_038677.3 | 4 | 589 | |||
| GenBank mRNA | AK144355.1 | - | 3 | 407 | ||
| BC032275.1 | - | 4 | 323 | 5370100 | ||
| L02210.1 | - | 4 | 564 | |||
| L21707.1 | - | 4 | 599 | |||
| L38394.1 | - | 4 | 512 | |||
| M98547.1 | - | 4 | 553 | |||

Ryk
Genomic Map