siRNA Details
| Gene Symbol: Khdrbs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011317.4 | NP_035447.3 | 9 | 1460 | ||
| GenBank mRNA | AK044927.1 | - | 9 | 1457 | ||
| AK050520.1 | - | 9 | 1455 | |||
| AK146815.1 | - | 9 | 1422 | |||
| AK150687.1 | - | 9 | 1439 | |||
| AK152084.1 | - | 9 | 1439 | |||
| BC002051.1 | - | 9 | 1416 | 3490667 | ||
| EL606063.1 | - | 5 | 735 | |||
| U17046.1 | - | 9 | 1417 | |||
| U17961.1 | - | 9 | 1428 | |||

Khdrbs1
Genomic Map