siRNA Details
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013651.4 | NP_038679.3 | 7 | 635 | ||
GenBank mRNA | BC052697.1 | - | 7 | 604 | 5715282 | |
CX214888.1 | - | 2 | 141 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013651.4 | NP_038679.3 | 7 | 635 | ||
GenBank mRNA | BC052697.1 | - | 7 | 604 | 5715282 | |
CX214888.1 | - | 2 | 141 |