siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204979.1 | NP_001191908.1 | 5 | 586 | ||
| NM_011319.3 | NP_035449.1 | 5 | 586 | |||
| GenBank mRNA | AK028358.1 | - | 5 | 518 | ||
| AK075827.1 | - | 5 | 517 | |||
| AK077309.1 | - | 5 | 468 | |||
| AK082793.1 | - | 5 | 497 | |||
| AK152150.1 | - | 5 | 544 | |||
| AK153158.1 | - | 5 | 536 | |||
| BC008612.1 | - | 5 | 475 | 3257101 | ||
| BC031932.1 | - | 5 | 475 | |||
| M74012.1 | - | 5 | 538 | |||

Sars
Genomic Map