siRNA Details
| Gene Symbol: Atxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199304.1 | NP_001186233.1 | 6 | 1759 | ||
| NM_001199305.1 | NP_001186234.1 | 7 | 1884 | |||
| NM_009124.6 | NP_033150.2 | 7 | 1884 | |||
| GenBank mRNA | AK048268.1 | - | 7 | 1871 | ||
| AK169931.1 | - | 7 | 1879 | |||
| BC058178.1 | - | 6 | 1725 | 5326559 | ||
| X83542.1 | - | 6 | 1625 | |||

Atxn1
Genomic Map