siRNA Details
| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077499.2 | NP_001070967.1 | 17 | 3234 | ||
| NM_011323.3 | NP_035453.2 | 16 | 3135 | |||
| GenBank mRNA | AF049617.1 | - | 16 | 3081 | ||
| AK083220.1 | - | 3 | 732 | |||
| U26707.1 | - | 14 | 2355 | |||

Scn8a
Genomic Map