siRNA Details
| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077499.2 | NP_001070967.1 | 14 | 2442 | ||
| NM_011323.3 | NP_035453.2 | 13 | 2343 | |||
| GenBank mRNA | AF049617.1 | - | 13 | 2289 | ||
| AK158609.1 | - | 14 | 2404 | |||
| U26707.1 | - | 11 | 1563 | |||

Scn8a
Genomic Map