siRNA Details
| Gene Symbol: Scnn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272023.1 | NP_001258952.1 | 8, 9 | 1376 | ||
| NM_011325.2 | NP_035455.1 | 8, 9 | 1373 | |||
| GenBank mRNA | AF112186.1 | - | 8, 9 | 1300 | ||
| AK028909.1 | - | 8, 9 | 1355 | |||
| AK036819.1 | - | 8, 9 | 1374 | |||
| AK087055.1 | - | 8, 9 | 1335 | |||
| AK164709.1 | - | 8, 9 | 1336 | |||
| BC131969.1 | - | 8, 9 | 1289 | 40130246 | ||
| BC132397.1 | - | 8, 9 | 1289 | 40130674 | ||

Scnn1b
Genomic Map