siRNA Details
| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011326.2 | NP_035456.1 | 11 | 1551 | ||
| GenBank mRNA | AF112187.1 | - | 11 | 1551 | ||
| AK143995.1 | - | 10 | 1244 | |||
| BC021338.1 | - | 11 | 1590 | 5042876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scnn1g| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011326.2 | NP_035456.1 | 11 | 1551 | ||
| GenBank mRNA | AF112187.1 | - | 11 | 1551 | ||
| AK143995.1 | - | 10 | 1244 | |||
| BC021338.1 | - | 11 | 1590 | 5042876 | ||