siRNA Details
| Gene Symbol: Scnn1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011326.2 | NP_035456.1 | 5, 6 | 1010 | ||
| GenBank mRNA | AF112187.1 | - | 5, 6 | 1010 | ||
| AK143995.1 | - | 4, 5 | 703 | |||
| BC021338.1 | - | 5, 6 | 1049 | 5042876 | ||

Scnn1g
Genomic Map