siRNA Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011327.4 | NP_035457.1 | 6 | 601 | ||
| GenBank mRNA | AK004860.2 | - | 6 | 531 | ||
| BC018384.1 | - | 6 | 556 | 3710340 | ||
| BC034613.1 | - | 6 | 522 | 5123611 | ||
| M91458.1 | - | 6 | 516 | |||

Scp2
Genomic Map