siRNA Details
| Gene Symbol: Cx3cl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009142.3 | NP_033168.2 | 3 | 1120 | ||
| GenBank mRNA | AF010586.1 | - | 3 | 1074 | ||
| AF071549.1 | - | 3 | 1111 | |||
| AK040220.1 | - | 3 | 1065 | |||
| AK138214.1 | - | 3 | 1097 | |||
| AK138472.1 | - | 3 | 1097 | |||
| AK138574.1 | - | 3 | 1097 | |||
| AK154646.1 | - | 3 | 1097 | |||
| BC006650.1 | - | 3 | 1074 | 3498747 | ||
| BC054838.1 | - | 3 | 1080 | 6490082 | ||
| U92565.1 | - | 3 | 1035 | |||

Cx3cl1
Genomic Map