siRNA Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011340.3 | NP_035470.3 | 8 | 1319 | ||
| GenBank mRNA | AA111321.1 | - | 1 | 123 | ||
| AF017057.1 | - | 8 | 1217 | |||
| AF036164.1 | - | 8 | 1217 | |||
| BC019852.1 | - | 8 | 1229 | 5123884 | ||
| D50460.1 | - | 8 | 1288 | |||
| D87975.1 | - | 8 | 1216 | |||

Serpinf1
Genomic Map