siRNA Details
| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011342.4 | NP_035472.1 | 4 | 520 | ||
| GenBank mRNA | AK010363.1 | - | 4 | 529 | ||
| AK017237.1 | - | 4 | 526 | |||
| AK034973.1 | - | 4 | 532 | |||
| AK037918.1 | - | 4 | 521 | |||
| AK089928.1 | - | 4 | 513 | |||
| AK134215.1 | - | 4 | 521 | |||
| AK190268.1 | - | 3 | 320 | |||
| BC009024.1 | - | 4 | 482 | 3481731 | ||
| U91538.1 | - | 4 | 477 | |||

Sec22b
Genomic Map