siRNA Details
| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039089.1 | NP_001034178.1 | 13 | 1418 | ||
| NM_011344.2 | NP_035474.1 | 13 | 1268 | |||
| GenBank mRNA | AF063095.1 | - | 13 | 1217 | ||
| AK005023.2 | - | 13 | 1391 | |||
| AK146022.1 | - | 13 | 1392 | |||
| AK160936.1 | - | 13 | 1226 | |||
| AK220502.1 | - | 13 | 1418 | |||
| BC026816.1 | - | 13 | 1240 | 4457637 | ||
| BC049959.1 | - | 8 | 630 | |||

Sel1l
Genomic Map