siRNA Details
| Gene Symbol: Sel1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039089.1 | NP_001034178.1 | 18 | 1928 | ||
| NM_011344.2 | NP_035474.1 | 18 | 1778 | |||
| GenBank mRNA | AF063095.1 | - | 18 | 1727 | ||
| AK005023.2 | - | 18 | 1901 | |||
| AK146022.1 | - | 18 | 1902 | |||
| AK160936.1 | - | 18 | 1736 | |||
| AK220502.1 | - | 18 | 1928 | |||
| BC026816.1 | - | 18 | 1750 | 4457637 | ||
| BC049959.1 | - | 13 | 1140 | |||
| BC057452.1 | - | 4 | 363 | |||

Sel1l
Genomic Map