siRNA Details
| Gene Symbol: Selp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011347.2 | NP_035477.1 | 13 | 2313 | ||
| GenBank mRNA | AK136243.1 | - | 4 | 681 | ||
| AK145335.1 | - | 1 | 283 | |||
| BC109158.1 | - | 12 | 2199 | 40047544 | ||
| BC109159.1 | - | 12 | 2199 | 40047547 | ||
| M72332.1 | - | 13 | 2313 | |||
| M87861.1 | - | 12 | 2199 | |||

Selp
Genomic Map