siRNA Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163489.1 | NP_001156961.1 | 13, 14 | 1652 | ||
| NM_001163490.1 | NP_001156962.1 | 13, 14 | 1731 | |||
| NM_001163491.1 | NP_001156963.1 | 13, 14 | 1690 | |||
| NM_013658.3 | NP_038686.3 | 13, 14 | 1830 | |||
| GenBank mRNA | AK035918.1 | - | 12, 13 | 1692 | ||
| AK077976.1 | - | 13, 14 | 1811 | |||
| AK152728.1 | - | 13, 14 | 1822 | |||
| AK153164.1 | - | 13, 14 | 1822 | |||
| AK154392.1 | - | 13, 14 | 1791 | |||
| BC025800.1 | - | 13, 14 | 1748 | 5102015 | ||
| X85991.1 | - | 13, 14 | 1652 | |||

Sema4a
Genomic Map