siRNA Details
| Gene Symbol: Sema4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163489.1 | NP_001156961.1 | 12 | 1469 | ||
| NM_001163490.1 | NP_001156962.1 | 12 | 1548 | |||
| NM_001163491.1 | NP_001156963.1 | 12 | 1507 | |||
| NM_013658.3 | NP_038686.3 | 12 | 1647 | |||
| GenBank mRNA | AK035918.1 | - | 11 | 1509 | ||
| AK077976.1 | - | 12 | 1628 | |||
| AK152728.1 | - | 12 | 1639 | |||
| AK153164.1 | - | 12 | 1639 | |||
| AK154392.1 | - | 12 | 1608 | |||
| BC025800.1 | - | 12 | 1565 | 5102015 | ||
| X85991.1 | - | 12 | 1469 | |||

Sema4a
Genomic Map