siRNA Details
| Gene Symbol: Sema4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281880.1 | NP_001268809.1 | 9 | 1123 | ||
| NM_013660.4 | NP_038688.2 | 8 | 1059 | |||
| GenBank mRNA | AK088653.1 | - | 5 | 341 | ||
| AK132688.1 | - | 9 | 1049 | |||
| AK150911.1 | - | 7 | 951 | |||
| BC049780.1 | - | 8 | 956 | 6509473 | ||
| U69535.1 | - | 6 | 586 | |||

Sema4d
Genomic Map