siRNA Details
| Gene Symbol: Sema5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009154.2 | NP_033180.2 | 7, 8 | 1080 | ||
| GenBank mRNA | AK031231.1 | - | 7, 8 | 1073 | ||
| AK043386.1 | - | 7, 8 | 792 | |||
| AK045236.1 | - | 7, 8 | 1084 | |||
| AK046502.1 | - | 7, 8 | 856 | |||
| AK141612.1 | - | 7, 8 | 1024 | |||
| AK143026.1 | - | 7, 8 | 1091 | |||
| AK157706.1 | - | 7, 8 | 1063 | |||
| BC065137.1 | - | 6, 7 | 549 | 30548165 | ||
| X97817.1 | - | 5, 6 | 449 | |||

Sema5a
Genomic Map