siRNA Details
| Gene Symbol: Sema5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009154.2 | NP_033180.2 | 12 | 2060 | ||
| GenBank mRNA | AK031231.1 | - | 12 | 2054 | ||
| AK043386.1 | - | 12 | 1772 | |||
| AK046502.1 | - | 12 | 1836 | |||
| AK077021.1 | - | 2 | 514 | |||
| AK141612.1 | - | 12 | 2004 | |||
| AK143026.1 | - | 12 | 2071 | |||
| AK157706.1 | - | 12 | 2043 | |||
| BC065137.1 | - | 11 | 1529 | 30548165 | ||
| X97817.1 | - | 10 | 1429 | |||

Sema5a
Genomic Map