siRNA Details
| Gene Symbol: Sema6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018744.2 | NP_061214.2 | 14 | 2168 | ||
| GenBank mRNA | AF030430.1 | - | 14 | 1572 | ||
| AF288666.1 | - | 14 | 1421 | |||
| AK042751.1 | - | 14 | 2169 | |||
| AK082711.1 | - | 14 | 2171 | |||
| BC059238.1 | - | 14 | 2130 | 6417475 | ||
| BC062979.1 | - | 14 | 1746 | 6841689 | ||

Sema6a
Genomic Map