siRNA Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042613.1 | NP_001036078.1 | 4 | 549 | ||
| NM_001042614.1 | NP_001036079.1 | 4 | 632 | |||
| NM_009155.3 | NP_033181.3 | 4 | 566 | |||
| GenBank mRNA | AK002450.1 | - | 4 | 548 | ||
| AK077265.1 | - | 4 | 548 | |||
| AK146774.1 | - | 4 | 548 | |||
| AK148623.1 | - | 4 | 548 | |||
| AK159269.1 | - | 4 | 548 | |||
| AK159524.1 | - | 4 | 549 | |||
| AK159894.1 | - | 4 | 545 | |||
| AK159911.1 | - | 4 | 549 | |||
| AK159935.1 | - | 4 | 548 | |||
| AK167611.1 | - | 4 | 548 | |||
| AK168463.1 | - | 4 | 545 | |||
| AK168583.1 | - | 4 | 545 | |||
| AK168631.1 | - | 4 | 548 | |||
| AK168747.1 | - | 4 | 548 | |||
| AK168926.1 | - | 4 | 548 | |||
| AK169011.1 | - | 4 | 547 | |||
| AK169158.1 | - | 4 | 547 | |||
| AK169362.1 | - | 4 | 548 | |||
| AK192055.1 | - | 3 | 383 | |||
| AK201439.1 | - | 3 | 388 | |||
| AK201752.1 | - | 3 | 383 | |||
| AK202422.1 | - | 3 | 384 | |||
| AK203751.1 | - | 3 | 383 | |||
| BC001991.1 | - | 4 | 549 | 3486761 | ||
| CB231357.1 | - | 4 | 569 | |||
| CK627189.1 | - | 4 | 549 | |||
| X99807.1 | - | 3 | 609 | |||

Sepp1
Genomic Map