siRNA Details
| Gene Symbol: Itsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198968.2 | NP_001185897.1 | 6 | 654 | ||
| NM_001198969.2 | NP_001185898.1 | 6 | 654 | |||
| NM_011365.4 | NP_035495.2 | 6 | 654 | |||
| GenBank mRNA | AF132479.1 | - | 5 | 394 | ||
| AF132480.1 | - | 5 | 394 | |||
| AK029325.1 | - | 6 | 601 | |||
| AK034367.1 | - | 6 | 655 | |||
| AK042449.1 | - | 6 | 624 | |||
| AK122480.1 | - | 1 | 41 | |||
| AK163807.1 | - | 6 | 628 | |||
| BC138262.1 | - | 6 | 631 | 8861281 | ||
| BC171950.1 | - | 6 | 631 | 9054236 | ||

Itsn2
Genomic Map