siRNA Details
| Gene Symbol: Sh3gl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277954.1 | NP_001264883.1 | 6 | 671 | ||
| NM_001277955.1 | NP_001264884.1 | 6 | 479 | |||
| NM_017400.6 | NP_059096.3 | 6 | 678 | |||
| GenBank mRNA | AK012114.1 | - | 421 | |||
| BC027096.1 | - | 678 | 5254139 | |||
| BC100453.1 | - | 516 | 6770826 | |||
| U58887.1 | - | 658 | ||||

Sh3gl3
Genomic Map