siRNA Details
| Gene Symbol: Sh3gl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277954.1 | NP_001264883.1 | 3 | 312 | ||
| NM_001277955.1 | NP_001264884.1 | 3 | 312 | |||
| NM_017400.6 | NP_059096.3 | 3 | 312 | |||
| GenBank mRNA | AK012114.1 | - | 254 | |||
| AK157464.1 | - | 266 | ||||
| BC027096.1 | - | 312 | 5254139 | |||
| BC100453.1 | - | 157 | 6770826 | |||
| BE653420.1 | - | 164 | ||||
| U58887.1 | - | 292 | ||||

Sh3gl3
Genomic Map