siRNA Details
| Gene Symbol: Shc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009167.3 | NP_033193.2 | 7 | 546 | ||
| GenBank mRNA | BC103611.1 | - | 7 | 546 | 40046304 | |
| BC103612.1 | - | 7 | 546 | 40046306 | ||
| BC105644.1 | - | 7 | 546 | 40046301 | ||
| BC105645.1 | - | 7 | 546 | 40046303 | ||
| U46854.1 | - | 7 | 546 | |||

Shc3
Genomic Map