siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 6 | 850 | ||
| GenBank mRNA | AK004613.1 | - | 6 | 851 | ||
| AK136381.1 | - | 853 | ||||
| BC003276.1 | - | 6 | 1752 | |||
| BC010208.1 | - | 802 | ||||
| X93999.1 | - | 6 | 814 | |||

St6galnac2
Genomic Map