siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 7, 8 | 964 | ||
| GenBank mRNA | AK004613.1 | - | 7, 8 | 965 | ||
| AK136381.1 | - | 967 | ||||
| BC003276.1 | - | 7, 8 | 1866 | |||
| BC010208.1 | - | 916 | ||||
| BC023076.1 | - | 1, 2 | 1412 | |||
| EU937514.1 | - | 1, 2 | 1411 | |||
| X93999.1 | - | 7, 8 | 928 | |||

St6galnac2
Genomic Map