siRNA Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009180.3 | NP_033206.2 | 5 | 680 | ||
| GenBank mRNA | AB027198.1 | - | 2 | 138 | ||
| AK004613.1 | - | 5 | 681 | |||
| AK136381.1 | - | 683 | ||||
| BC003276.1 | - | 5 | 1582 | |||
| BC010208.1 | - | 632 | ||||
| X93999.1 | - | 5 | 644 | |||

St6galnac2
Genomic Map