siRNA Details
| Gene Symbol: St6galnac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011372.2 | NP_035502.1 | 3 | 554 | ||
| GenBank mRNA | AK034804.1 | - | 3 | 556 | ||
| BC058387.1 | - | 3 | 539 | 6817021 | ||
| BC131987.1 | - | 3 | 505 | 40130264 | ||
| BC132317.1 | - | 3 | 505 | 40130594 | ||
| Y11342.1 | - | 3 | 554 | |||

St6galnac3
Genomic Map