siRNA Details
| Gene Symbol: Sim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011376.3 | NP_035506.2 | 6, 7 | 1000 | ||
| GenBank mRNA | AB221630.1 | - | 6, 7 | 754 | ||
| D79209.1 | - | 6, 7 | 793 | |||
| U40575.1 | - | 6, 7 | 997 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sim1| Gene Symbol: Sim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011376.3 | NP_035506.2 | 6, 7 | 1000 | ||
| GenBank mRNA | AB221630.1 | - | 6, 7 | 754 | ||
| D79209.1 | - | 6, 7 | 793 | |||
| U40575.1 | - | 6, 7 | 997 | |||