siRNA Details
| Gene Symbol: Ski | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011385.2 | NP_035515.2 | 2 | 1055 | ||
| GenBank mRNA | AF435852.1 | - | 2 | 1034 | ||
| AK019148.2 | - | 2 | 112 | |||
| BC068305.1 | - | 3 | 899 | 30093626 | ||
| CA451127.1 | - | 2 | 379 | |||

Ski
Genomic Map