siRNA Details
| Gene Symbol: Ski | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011385.2 | NP_035515.2 | 5 | 1538 | ||
| GenBank mRNA | AF435852.1 | - | 5 | 1517 | ||
| AK019148.2 | - | 5 | 595 | |||
| BC030841.1 | - | 2 | 315 | |||
| BC068305.1 | - | 6 | 1382 | 30093626 | ||
| CB722801.1 | - | 2 | 247 | |||

Ski
Genomic Map