siRNA Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205311.1 | NP_001192240.1 | 7 | 888 | ||
| NM_019415.2 | NP_062288.2 | 7 | 885 | |||
| GenBank mRNA | AK052691.1 | - | 7 | 889 | ||
| AK052755.1 | - | 7 | 887 | |||
| AK085496.1 | - | 7 | 892 | |||
| BC038612.1 | - | 7 | 910 | 4237274 | ||
| U61085.1 | - | 7 | 867 | |||

Slc12a3
Genomic Map