siRNA Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022411.3 | NP_071856.1 | 7, 8 | 1105 | ||
| GenBank mRNA | AF201903.1 | - | 7, 8 | 1105 | ||
| AK140972.1 | - | 7, 8 | 1156 | |||
| BC013493.1 | - | 7, 8 | 1130 | 4218337 | ||

Slc13a2
Genomic Map