siRNA Details
| Gene Symbol: Slc16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009196.4 | NP_033222.1 | 3 | 513 | ||
| GenBank mRNA | AF058055.1 | - | 3 | 494 | ||
| AK033516.1 | - | 3 | 511 | |||
| AK079379.1 | - | 3 | 509 | |||
| AK088762.1 | - | 3 | 511 | |||
| AK161701.1 | - | 3 | 511 | |||
| AK162682.1 | - | 3 | 510 | |||
| AK166606.1 | - | 3 | 511 | |||
| BC014777.1 | - | 3 | 521 | 3499773 | ||
| X82438.1 | - | 2 | 305 | |||

Slc16a1
Genomic Map