siRNA Details
| Gene Symbol: Slc16a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009197.2 | NP_033223.2 | 1 | 558 | ||
| GenBank mRNA | AB265788.1 | - | 1 | 384 | ||
| AF045692.1 | - | 1 | 606 | |||
| BC080678.1 | - | 1 | 394 | 6313495 | ||
| BC096587.1 | - | 1 | 494 | 3992886 | ||
| BM935188.1 | - | 1 | 71 | |||
| CX218841.1 | - | 1 | 494 | |||

Slc16a2
Genomic Map