siRNA Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159593.1 | NP_001153065.1 | 4, 5 | 1067 | ||
| NM_015747.2 | NP_056562.1 | 4, 5 | 1067 | |||
| GenBank mRNA | AK035898.1 | - | 4, 5 | 1038 | ||
| AK155506.1 | - | 4, 5 | 1040 | |||
| BC015085.1 | - | 4, 5 | 967 | 4015575 | ||
| M73696.1 | - | 3, 4 | 986 | |||

Slc20a1
Genomic Map