siRNA Details
| Gene Symbol: Slc22a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011395.2 | NP_035525.1 | 6 | 1409 | ||
| GenBank mRNA | AF078750.1 | - | 6 | 1409 | ||
| AF082566.1 | - | 6 | 1292 | |||
| AK143988.1 | - | 6 | 1079 | |||
| BC137661.1 | - | 6 | 1284 | 8860680 | ||
| BC137662.1 | - | 6 | 1284 | 8860681 | ||

Slc22a3
Genomic Map