siRNA Details
| Gene Symbol: Slc2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031197.2 | NP_112474.2 | 10 | 1375 | ||
| GenBank mRNA | AK005068.1 | - | 10 | 1376 | ||
| AK149532.1 | - | 3 | 488 | |||
| AK165430.1 | - | 10 | 1310 | |||
| BC034675.1 | - | 10 | 1294 | 4236331 | ||
| S77926.1 | - | 2 | 114 | |||
| X15684.1 | - | 10 | 1324 | |||
| X16986.1 | - | 10 | 1314 | |||

Slc2a2
Genomic Map