siRNA Details
| Gene Symbol: Slc2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011401.4 | NP_035531.3 | 9, 10 | 1635 | ||
| GenBank mRNA | AK028041.1 | - | 9, 10 | 1411 | ||
| AK045328.1 | - | 9, 10 | 1411 | |||
| AK147258.1 | - | 8, 9 | 3947 | |||
| AK155997.1 | - | 9, 10 | 1394 | |||
| AK164286.1 | - | 9, 10 | 1407 | |||
| BC034122.1 | - | 9, 10 | 1393 | 4527667 | ||
| BC050095.1 | - | 9, 10 | 1389 | |||
| BC058811.1 | - | 9, 10 | 1393 | 6490844 | ||
| CT010354.1 | - | 8, 9 | 1280 | |||
| M75135.1 | - | 9, 10 | 1359 | |||
| X61093.1 | - | 9, 10 | 1359 | |||

Slc2a3
Genomic Map