siRNA Details
| Gene Symbol: Slc34a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011402.3 | NP_035532.2 | 10 | 1234 | ||
| GenBank mRNA | AF081499.1 | - | 10 | 1231 | ||
| AK004832.1 | - | 10 | 1233 | |||
| AK145599.1 | - | 10 | 1233 | |||
| BC096369.1 | - | 9 | 1205 | 3962784 | ||

Slc34a2
Genomic Map