siRNA Details
| Gene Symbol: Slc3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009205.2 | NP_033231.2 | 7 | 1342 | ||
| GenBank mRNA | AK052623.1 | - | 7 | 1313 | ||
| AK132275.1 | - | 7 | 1314 | |||
| BC013441.1 | - | 7 | 1284 | 4222972 | ||
| D88533.1 | - | 7 | 1302 | |||

Slc3a1
Genomic Map