siRNA Details
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253892.1 | NP_001240821.1 | 12 | 1875 | ||
NM_009207.3 | NP_033233.2 | 13 | 2071 | |||
GenBank mRNA | AK145930.1 | - | 13 | 1996 | ||
AK147858.1 | - | 13 | 2027 | |||
BC054102.1 | - | 12 | 1880 | 30017310 | ||
J04036.1 | - | 13 | 1986 |
