siRNA Details
| Gene Symbol: Slc4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253892.1 | NP_001240821.1 | 3 | 498 | ||
| NM_009207.3 | NP_033233.2 | 4 | 694 | |||
| GenBank mRNA | AK145930.1 | - | 4 | 619 | ||
| AK147858.1 | - | 4 | 650 | |||
| AK212015.1 | - | 1 | 151 | |||
| BC054102.1 | - | 3 | 503 | 30017310 | ||
| J04036.1 | - | 4 | 609 | |||

Slc4a2
Genomic Map